Learn More
National Safety Apparel™ 65 Cal ArcGuard™ Nomex/Kevlar Lift Front Hood
SDP
Catalog No. 17001721 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Caramel
Catalog No.
17-001-721 Color: Caramel
1 options
Catalog No. 17-001-721 Supplier National Safety Apparel™ Supplier No. H65KDQT65LF
Inherently flame-resistant so they won’t burn, support combustion, or melt
  • Arc Rating: 65 cal/cm²
  • CAT 4\
  • Fabric/Material: Multilayer DuPont™ Nomex™ and Kevlar™
  • Hard hat included
  • Knit gussets at sides provide greater range of motion and flexibility
  • Lift front shield makes it easy to obtain fresh air while out of the hazard risk zone
  • Premium anti-fog and scratch resistant coating on faceshield
  • PureView™ faceshield technology features a greater range of visibility
  • Made in the USA
Specifications
Size Medium
Material DuPont™ Nomex™, Kevlar™
Color Caramel
Closure Type Snap
Seam Type Stitched
Arc Flash Rating 65 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F2178, OSHA 1910.269