Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Calibration Standard, 650 mg/L Phosphate, Ion Chromatography, NIST Traceable

Catalog No. 502392616 Shop All Waters Corporation Products
Encompass_Preferred

This 650 mg/L Phosphate Calibration Standard is for ion chromatography, used to analyze phosphate/phosphoric acid levels in beverages. It contains 650 mg/L potassium phosphate monohydrate and 10 mg/L potassium sulfate in ultra-pure deionized water. Directly traceable to NIST Reference Standards and meeting ISO Guide 31, each shipment includes a Certificate of Analysis.

  • Separation mode: Ion chromatography
  • Detection method: Conductivity
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC
  • Quantitative: Yes
  • Shelf life: 12 months
  • Traceability: NIST
  • Shipping condition: Ambient
  • Volume: 125 mL
  • Type: Calibration
  • Application: System calibration
  • Product type: Standards
  • Units per package: 1 pk

Catalog No. 50-239-2616 Supplier Waters Corp Supplier No. 186008050
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 650 mg/L Phosphate Calibration Standard is for ion chromatography, used to analyze phosphate/phosphoric acid levels in beverages. It contains 650 mg/L potassium phosphate monohydrate and 10 mg/L potassium sulfate in ultra-pure deionized water. Directly traceable to NIST Reference Standards and meeting ISO Guide 31, each shipment includes a Certificate of Analysis.

  • Separation mode: Ion chromatography
  • Detection method: Conductivity
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: LC
  • Quantitative: Yes
  • Shelf life: 12 months
  • Traceability: NIST
  • Shipping condition: Ambient
  • Volume: 125 mL
  • Type: Calibration
  • Application: System calibration
  • Product type: Standards
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.