Learn More
Ace Glass, Inc. Combo mantle, 3 position, low temperature, fits 650mL round bottom flasks, max diameter 4.25 inches, 240v 200 watts per position, order code (-63) or (-73) power supply separately
SDP
Catalog No. 50977366
Encompass
Combo mantle, 3 position, low temperature, fits 650mL round bottom flasks, max diameter 4.25 inches, 240v 200 watts per position, order code (-63) or (-73) power supply separately
Catalog No. 50-977-366 Supplier Ace Glass, Inc. Supplier No. 12061134
Combo mantle, 3 position, low temperature, fits 650mL round bottom flasks, max diameter 4.25 inches, 240v 200 watts per position, order code (-63) or (-73) power supply separately