Learn More
Surmodics Inc 650 nm Liquid Stop Solution for TMB Microwell Substrates, 1L
SDP
Catalog No. NC0561125
Encompass_Preferred
A liquid stop solution used in end point ELISA applications where a TMB substrate is used as the detection mechanism. Absorbance is read in the blue spectral range (650 nm).
Catalog No. NC0561125 Supplier Surmodics Inc Supplier No. LBSP100001
A liquid stop solution used in end point ELISA applications where a TMB substrate is used as the detection mechanism. Absorbance is read in the blue spectral range (650 nm).