Learn More
Surmodics Inc 650 nm Stop Reagent for TMB Microwell Substrates, 1L
SDP
Catalog No. NC0217060
Encompass_Preferred
The BioFX™ 650 nm Stop Reagent for TMB Microwell Substrates (1 L) is a proprietary dry blend used to stop TMB microwell substrate reactions without causing a color or absorbance change. Absorbance is read in the blue spectral range (650 nm)
Catalog No. NC0217060 Supplier Surmodics Inc Supplier No. BSTP100001
The BioFX™ 650 nm Stop Reagent for TMB Microwell Substrates (1 L) is a proprietary dry blend used to stop TMB microwell substrate reactions without causing a color or absorbance change. Absorbance is read in the blue spectral range (650 nm)