Learn More
Sino Biological Recombinant Rabbit NGFR/p75NTR Protein (ECD, hFc Tag)
Supplier:  Sino Biological 65160T02H50
Encompass_Preferred
A DNA sequence encoding the rabbit NGFR (XP_008269543.1) (Met1-Asp242) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3262