Learn More
TARGETMOL CHEMICALS INC 653-47 hydrochloride 5MG
SDP
Catalog No. 502876482
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 653-47 hydrochloride is a potentiator, significantly potentiates the cAMP-response element binding protein (CREB) inhibitory activity of 666-15. It is also a very weak CREB inhibitor with IC50 of 26.3 uM. Purity 99.65%
Catalog No. 50-287-6482 Supplier TARGETMOL CHEMICALS INC Supplier No. T88905MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 653-47 hydrochloride is a potentiator, significantly potentiates the cAMP-response element binding protein (CREB) inhibitory activity of 666-15. It is also a very weak CREB inhibitor with IC50 of 26.3 uM. Purity 99.65%