Learn More
Medchemexpress LLC HY-134598A 5mg Medchemexpress, 653-47 (hydrochloride) CAS:1224567-46-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134598A5MG
Encompass_Preferred
Medchemexpress, HY-134598A 5mg 653-47 (hydrochloride) CAS:1224567-46-7 653-47 hydrochloride, a potentiator, significantly potentiates the cAMP-response element binding protein (CREB) inhibitory activity of 666-15. 653-47 hydrochloride is also a very weak CREB inhibitor with IC50 of 26.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1371