Learn More
NEOGEN Petrifilm Salmonella Express Plates, 50 Plates
SDP
Catalog No. NC2874619
Encompass_Preferred
The plate offers presumptive positive results for Salmonella spp. in just 40 hours for processed foods and 48 hours for unprocessed foods. Confirmation disc easily added to plate for biochemical confirmation, results in 4-5 hours. Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes. Only one enrichment for low-background, processed foods.
Catalog No. NC2874619 Supplier NEOGEN Supplier No. 700002144
The plate offers presumptive positive results for Salmonella spp. in just 40 hours for processed foods and 48 hours for unprocessed foods. Confirmation disc easily added to plate for biochemical confirmation, results in 4-5 hours. Plates are sample-ready, eliminating all the time-consuming steps of preparing media/agar dishes. Only one enrichment for low-background, processed foods.