Learn More
Caron Products And Services PM Kit Model 6540 VIS Lamps 230V-60Hz
Catalog No. NC2331835
Encompass_Preferred
Preventative Maintenance Kit for Model 6540-2 (230V-60Hz Photostability Chamber) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing, (14) LGT-139 VIS, Cool White Lamps, and (4) VEL-104 1/2" Velcro Discs. Detailed Instructions Included.
Catalog No. NC2331835 Supplier Caron Products And Services Supplier No. PM65402VIS
Preventative Maintenance Kit for Model 6540-2 (230V-60Hz Photostability Chamber) Includes (1) FIL-114 Filter Media, 25 PPI, with Reinforced Backing, (14) LGT-139 VIS, Cool White Lamps, and (4) VEL-104 1/2" Velcro Discs. Detailed Instructions Included.