Learn More
Ace Glass, Inc. 65mm powder funnel with 19/38 inner joint
SDP
Catalog No. 50974706
Encompass_Preferred
65mm powder funnel with 19/38 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-974-706 Supplier Ace Glass, Inc. Supplier No. 723606
65mm powder funnel with 19/38 inner joint is useful in pouring powders or liquids into ground joint containers.