Learn More
Alkali Scientific ASI™ 65mm RODAC Dish, 20/Pk, 500/Cs
SDP
Catalog No. 503028996
Encompass_Preferred
The 65 mm RODAC (Replicate Organism Detection and Counting) dish is specifically designed for surface sampling and microbial monitoring. With a compact 65 mm diameter, this dish is ideal for use in quality control, environmental testing, and other microbial sampling applications. Manufactured from durable, optically clear plastic, it provides excellent visibility for accurate sample evaluation. Each dish is pre-sterilized, ensuring a contamination-free environment and ready-to-use convenience. Packaged 20 per pack and 500 per case, these dishes are a reliable and economical choice for laboratories and industries requiring consistent and precise microbial analysis.
Catalog No. 50-302-8996 Supplier Alkali Scientific Supplier No. RD0065
The 65 mm RODAC (Replicate Organism Detection and Counting) dish is specifically designed for surface sampling and microbial monitoring. With a compact 65 mm diameter, this dish is ideal for use in quality control, environmental testing, and other microbial sampling applications. Manufactured from durable, optically clear plastic, it provides excellent visibility for accurate sample evaluation. Each dish is pre-sterilized, ensuring a contamination-free environment and ready-to-use convenience. Packaged 20 per pack and 500 per case, these dishes are a reliable and economical choice for laboratories and industries requiring consistent and precise microbial analysis.