Learn More
Haztech Systems, Inc HW4072 Pyrex Watch Glass. 65 mm diameter.
SDP
Catalog No. NC9249855
Encompass
HW4072 Pyrex Watch Glass 65mm diameter. Watch glass has a heavy walled fire-polished edge to provide mechanical strength and high chemical durability.
Catalog No. NC9249855 Supplier Haztech Systems, Inc Supplier No. HW4072
HW4072 Pyrex Watch Glass 65mm diameter. Watch glass has a heavy walled fire-polished edge to provide mechanical strength and high chemical durability.