Learn More
Thomas Scientific FluidX Automation Friendly External Thread Cap, Orange, 24-format, 240/pk
SDP
Catalog No. NC1824603
Encompass_Preferred
Suitable for 4ml Tissue Tube, 6ml and 10ml PP Jacket Tubes. Specifically designed for long-term storage of tissue samples. FluidX Tri-Coded Jacket offers unequaled audit traceability, enabling data sharing and sample tracking between multiple labs, locations and automation capabilities.
Catalog No. NC1824603 Supplier Thomas Scientific Supplier No. 1150L93
Suitable for 4ml Tissue Tube, 6ml and 10ml PP Jacket Tubes. Specifically designed for long-term storage of tissue samples. FluidX Tri-Coded Jacket offers unequaled audit traceability, enabling data sharing and sample tracking between multiple labs, locations and automation capabilities.