Learn More
MSC Tape Logic Packing Tape, 3 in, Hot Melt, 110 yd
Catalog No. 501918523
Encompass
This packing tape is a cold temperature carton sealing tape with a pressure-sensitive hot melt adhesive. It is formulated for use in refrigerated or low-temperature environments and is chemically acceptable by the USDA for packaging materials with minimal contact with meat or poultry food products.
  • Designed for refrigerated or low-temperature environments
  • Pressure-sensitive hot melt adhesive
  • Chemically acceptable by USDA for minimal food contact
  • Made from polypropylene material
  • Measures 1.7 mil thick
  • Clear color
Catalog No. 50-191-8523 Supplier MSC Supplier No. 39581137
This packing tape is a cold temperature carton sealing tape with a pressure-sensitive hot melt adhesive. It is formulated for use in refrigerated or low-temperature environments and is chemically acceptable by the USDA for packaging materials with minimal contact with meat or poultry food products.
  • Designed for refrigerated or low-temperature environments
  • Pressure-sensitive hot melt adhesive
  • Chemically acceptable by USDA for minimal food contact
  • Made from polypropylene material
  • Measures 1.7 mil thick
  • Clear color