Learn More
Chemglass Life Sciences Dewar Flask, Cylindrical, Wide Mouth, 665mL Capacity, 70mm ID X 195mm Inside Depth, 86mm OD X 241mm Height
SDP
Catalog No. CG159302
Encompass_Preferred
Dewar Flask, Cylindrical, Wide Mouth, 665mL Capacity, 70mm ID X 195mm Inside Depth, 86mm OD X 241mm Height. Dewar flask with weighted metal base for stability and protective plastic mesh cover providing a firm grip and a cushion in the event the flask is bumped. Supplied complete with a vented polyethylene stopper to minimize evaporation.
Catalog No. CG159302 Supplier Chemglass Life Sciences Supplier No. CG159302
Dewar Flask, Cylindrical, Wide Mouth, 665mL Capacity, 70mm ID X 195mm Inside Depth, 86mm OD X 241mm Height. Dewar flask with weighted metal base for stability and protective plastic mesh cover providing a firm grip and a cushion in the event the flask is bumped. Supplied complete with a vented polyethylene stopper to minimize evaporation.