Learn More
Medchemexpress LLC 666-15 | 1433286-70-4 | MFCD30182343 | 99.8% | 620.52 g/mol | C33H31Cl2N3O5 | 1 MG
SDP
Catalog No. 5000213067
Encompass_Preferred
A potent, selective CREB inhibitor with an IC50 of 81 nM that has been reported to suppress tumor growth in breast cancer xenograft models. Supplied as a high-purity solid, the compound is provided for research use with defined storage and stability recommendations.
  • Potent, selective CREB inhibition with IC50 = 81 nM.
  • Demonstrated tumor growth suppression in breast cancer xenograft models.
  • High purity, approximately 99.8%.
  • Chemical formula C33H31Cl2N3O5; molecular weight 620.52 g/mol.
  • White to off-white solid; store solid at 4°C and store solutions at -80°C for long-term stability.
  • Supplied in milligram-scale quantities suitable for laboratory research.
Catalog No. 50-002-13067 Supplier Medchemexpress LLC Supplier No. HY1011201MG
A potent, selective CREB inhibitor with an IC50 of 81 nM that has been reported to suppress tumor growth in breast cancer xenograft models. Supplied as a high-purity solid, the compound is provided for research use with defined storage and stability recommendations.
  • Potent, selective CREB inhibition with IC50 = 81 nM.
  • Demonstrated tumor growth suppression in breast cancer xenograft models.
  • High purity, approximately 99.8%.
  • Chemical formula C33H31Cl2N3O5; molecular weight 620.52 g/mol.
  • White to off-white solid; store solid at 4°C and store solutions at -80°C for long-term stability.
  • Supplied in milligram-scale quantities suitable for laboratory research.