Learn More
Medchemexpress LLC 666-15 | 1433286-70-4 | MFCD30182343 | 99.8% | 620.52 g/mol | C33H31Cl2N3O5 | 1 ML
SDP
Catalog No. 5000213505
Encompass_Preferred
666-15 is a potent, selective small-molecule inhibitor of CREB-mediated transcription (IC50 = 81 nM) with demonstrated activity in cellular assays and preclinical tumor models. The compound is supplied as a ready-to-use DMSO solution and is provided with analytical documentation for experimental use.
  • Potent CREB inhibition (IC50 = 81 nM).
  • Demonstrated activity in cellular and animal models.
  • Supplied as a 10 mM solution in DMSO for convenience.
  • High purity confirmed by HPLC (>99.7%).
  • Provided with certificate of analysis and safety data.
Catalog No. 50-002-13505 Supplier Medchemexpress LLC Supplier No. HY10112010MM 1ML
666-15 is a potent, selective small-molecule inhibitor of CREB-mediated transcription (IC50 = 81 nM) with demonstrated activity in cellular assays and preclinical tumor models. The compound is supplied as a ready-to-use DMSO solution and is provided with analytical documentation for experimental use.
  • Potent CREB inhibition (IC50 = 81 nM).
  • Demonstrated activity in cellular and animal models.
  • Supplied as a 10 mM solution in DMSO for convenience.
  • High purity confirmed by HPLC (>99.7%).
  • Provided with certificate of analysis and safety data.