Learn More
MSC Perma-Patch Crack Filler, Asphaltic, Gray, 1 gal, Ultra-low Modulus
Supplier:  MSC 72194319
Encompass
This asphaltic, pourable, ultra-low modulus, neutral cure elastomeric emulsion is designed for effective crack sealing. It offers superior flexibility and resistance compared to other sealants, making it suitable for both new construction and remedial repairs in existing structures.
  • Single-component, ready-to-use, ultra-low modulus, pourable, neutral cure elastomeric emulsion.
  • Provides a long-lasting, flexible, and durable seal.
  • Easy, self-leveling installation.
  • Prevents water and noncompressibles from damaging pavement and foundation.
  • Effective where other joint sealing materials have failed.
  • Outstanding weathering resistance; flexible down to -20°F (-29°C).
  • Jet-blast resistant and serviceable against fuel/oil spills.
  • Bonds strongly to asphalt or concrete without primer.
  • Self-leveling properties eliminate the need for tooling.
Catalog No. 50-283-6787