Learn More
MSC Perma-Patch Crack Sealant, Gray, 55 Gallon, Cold-Pour
Catalog No. 502838320
Encompass
This single-component, ready-to-use cold-pour crack sealant is designed for pavement maintenance. It meets ASTM D6690 specifications, offering efficient and superior performance for asphalt and concrete pavements. Ideal for government road departments and paving contractors.
  • Meets ASTM D6690 specifications.
  • Requires less expensive equipment.
  • Enhances productivity, saving start-up and cleanup time.
  • Prevents pavement deterioration by sealing cracks.
  • Easy to use with manual/pump application; self-leveling.
  • Environmentally friendly; water cleanup for tools.
  • Increases safety; eliminates hot-applied sealant risks.
Catalog No. 50-283-8320 Supplier MSC Supplier No. 72194574
Add to Cart
Add to Cart
Request a Quote
This single-component, ready-to-use cold-pour crack sealant is designed for pavement maintenance. It meets ASTM D6690 specifications, offering efficient and superior performance for asphalt and concrete pavements. Ideal for government road departments and paving contractors.
  • Meets ASTM D6690 specifications.
  • Requires less expensive equipment.
  • Enhances productivity, saving start-up and cleanup time.
  • Prevents pavement deterioration by sealing cracks.
  • Easy to use with manual/pump application; self-leveling.
  • Environmentally friendly; water cleanup for tools.
  • Increases safety; eliminates hot-applied sealant risks.