Learn More
MSC Perma-Patch Crack Sealant, ASTM D6690 Compliant, Cold Pour, 1 Gallon
Catalog No. 502838546
Encompass
This single-component, ready-to-use cold pour crack sealant is designed for efficient pavement maintenance and sealing. It meets ASTM D6690 specifications, offering superior performance and operational efficiency for both new construction and repair applications.
  • Meets ASTM D6690 specifications as a cold pour solution.
  • Requires significantly less expensive equipment than hot melt systems.
  • Saves hours in start-up, operation, and cleanup, enhancing productivity.
  • Effectively seals cracks to prevent water infiltration and extend pavement life.
  • Self-leveling formula ensures thorough filling; bonds to asphalt or concrete without primer.
  • Remains flexible down to -20°F, jet-blast resistant, and open to traffic in 15 minutes.
  • Environmentally friendly with water cleanup for tools.
Catalog No. 50-283-8546 Supplier MSC Supplier No. 72193899
This single-component, ready-to-use cold pour crack sealant is designed for efficient pavement maintenance and sealing. It meets ASTM D6690 specifications, offering superior performance and operational efficiency for both new construction and repair applications.
  • Meets ASTM D6690 specifications as a cold pour solution.
  • Requires significantly less expensive equipment than hot melt systems.
  • Saves hours in start-up, operation, and cleanup, enhancing productivity.
  • Effectively seals cracks to prevent water infiltration and extend pavement life.
  • Self-leveling formula ensures thorough filling; bonds to asphalt or concrete without primer.
  • Remains flexible down to -20°F, jet-blast resistant, and open to traffic in 15 minutes.
  • Environmentally friendly with water cleanup for tools.