Learn More
MSC Perma-Patch Crack Sealant, Cold Pour, Gray, 1 Gallon
Catalog No. 502836784
Encompass
Perma-Patch 6690 ColdFuze Gray is a single-component, ready-to-use, cold-pour crack sealant for pavement maintenance. It is the first cold-pour solution to meet ASTM D6690 specifications, effectively sealing cracks to prevent deterioration and extend pavement life. Its self-leveling formula ensures thorough filling for new construction and repairs.
  • Meets ASTM D6690 and C920 standards.
  • Reduces equipment costs and boosts productivity.
  • Prevents pavement deterioration.
  • Self-leveling for easy, thorough filling.
  • Environmentally friendly: water cleanup, no solvents.
  • Applicable above 40°F, flexible to -20°F, jet-blast resistant.
  • Bonds to asphalt or concrete without primer.
  • Trafficable within 15 minutes.
Catalog No. 50-283-6784 Supplier MSC Supplier No. 72193709
Perma-Patch 6690 ColdFuze Gray is a single-component, ready-to-use, cold-pour crack sealant for pavement maintenance. It is the first cold-pour solution to meet ASTM D6690 specifications, effectively sealing cracks to prevent deterioration and extend pavement life. Its self-leveling formula ensures thorough filling for new construction and repairs.
  • Meets ASTM D6690 and C920 standards.
  • Reduces equipment costs and boosts productivity.
  • Prevents pavement deterioration.
  • Self-leveling for easy, thorough filling.
  • Environmentally friendly: water cleanup, no solvents.
  • Applicable above 40°F, flexible to -20°F, jet-blast resistant.
  • Bonds to asphalt or concrete without primer.
  • Trafficable within 15 minutes.