Learn More
Ace Glass, Inc. 6696 NO.25 PVC CTD ADAPTER 6696 NO.25 PVC CTD ADAPTER
SDP
Supplier:  Ace Glass, Inc. 669670
Encompass_Preferred
#25 Ace-Thred adapter for use with umbrella freeze drying apparatus utilizing Ace-Threds for positive, grease-free connections that are easy to manipulate.
Catalog No. 50-973-328