Learn More
MSC Permatex. 80050 3 oz Tube Clear RTV Silicone Joint Sealant -75 to 400°F Operating Temp, 24 hr Full Cure Time
Catalog No. 501113830
Encompass
Permatex. 80050 3 oz Tube Clear RTV Silicone Joint Sealant -75 to 400°F Operating Temp, 24 hr Full Cure Time
Catalog No. 50-111-3830 Supplier MSC Supplier No. 64478043
Permatex. 80050 3 oz Tube Clear RTV Silicone Joint Sealant -75 to 400°F Operating Temp, 24 hr Full Cure Time