Learn More
Sino Biological Recombinant Feline IL-10 Protein
Supplier:  Sino Biological 67001UNAE20
Encompass_Preferred
A DNA sequence encoding the feline IL10 (P55029.2) (Ser19-Ile178) was expressed with an initial Met at the N-terminal.
Catalog No. 50-162-3266