Learn More
Cole-Parmer 6781S GRINDING VIAL SET
Supplier:  Cole-Parmer 0457780
Encompass_Preferred
  • Designed for efficient, contamination-free cryogenic grinding.
  • Holds up to 4 mL with a sample capacity of 0.5 to 4 mL.
  • Stainless steel construction for durability.
  • Prevents cross-contamination and retains volatiles.
  • Compatible with Freezer/Mill cryogenic grinders.
  • Includes one stainless steel cylinder, two end plugs, an impactor, and 10 O-rings.
  • Suitable for grinding various materials like tissue, seeds, and polymers.
  • Reusable and autoclavable for convenient cleaning.
  • Protects hazardous and critical samples during grinding.
  • Easy to clean for better hygiene.
Catalog No. NC2822622