Learn More
Harrell Industries 68/32 30G PHENOL/METHANOL
SDP
Catalog No. NC3359479
Encompass
30 gallons of phenol/methanol mix.(68/32) Specifications:Moisture: 0.25% max.Phenol: 68.0% ± 0.5%Color (APHA):50 max.Specific Gravity @ 25 °C:0.9700 ± 0.003Blank Titration To pass test
Catalog No. NC3359479 Supplier Harrell Industries Supplier No. 618004
30 gallons of phenol/methanol mix.(68/32) Specifications:Moisture: 0.25% max.Phenol: 68.0% ± 0.5%Color (APHA):50 max.Specific Gravity @ 25 °C:0.9700 ± 0.003Blank Titration To pass test