Learn More
Abcam DyLight 680 Conjugation Kit (Fast) - Lightning-Link, 100UG
Catalog No. NC1969193
Encompass_Preferred
DyLight 680 conjugation / labeling in < 20 mins with 30 secs hands-on time using DyLight 680 Conjugation Kit (Fast) - Lightning-Link ab201804. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1969193 Supplier Abcam Supplier No. AB201804100UG
DyLight 680 conjugation / labeling in < 20 mins with 30 secs hands-on time using DyLight 680 Conjugation Kit (Fast) - Lightning-Link ab201804. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement