Learn More
eMolecules 683239-16-9 | 20-(tert-Butoxy)-20-oxoicosanoic acid | ChemScene | MFCD23136041 | 398.628 | C24H46O4 | 97.000 | CC(C)(C)OC(=O)CCCCCCCCCCCCCCCCCCC(O)=O | 25g | 582635189
Shop All eMolecules Inc. Products