Learn More
MSC CHEM RESIST/DISP COVERALL
Supplier:  MSC 32270472
Encompass
Disposable Coveralls: Chemical, Size 2X-Large, 2.9800 oz, Film Laminate, Storm Flap w/Double Zipper Closure
Catalog No. 50-273-9008