Learn More
Medchemexpress LLC BMS-986187 | 684238-37-7 | 5 MG
SDP
Catalog No. 5000231911
Encompass_Preferred
BMS-986187 is a small-molecule positive allosteric modulator selective for the δ-opioid receptor, characterized for potentiation of receptor signaling and used in studies of signaling bias.
  • Potent δ-opioid receptor PAM with reported EC50 ≈ 0.03 μM.
  • High purity solid, supplied as white to off-white powder.
  • Available in research quantities such as 5 mg and as a 10 mM solution in DMSO.
  • Molecular weight 470.6 g/mol; chemical formula C31H34O4.
  • Suitable for in vitro pharmacology and receptor mechanism studies.
Catalog No. 50-002-31911 Supplier Medchemexpress LLC Supplier No. HY1206135MG
BMS-986187 is a small-molecule positive allosteric modulator selective for the δ-opioid receptor, characterized for potentiation of receptor signaling and used in studies of signaling bias.
  • Potent δ-opioid receptor PAM with reported EC50 ≈ 0.03 μM.
  • High purity solid, supplied as white to off-white powder.
  • Available in research quantities such as 5 mg and as a 10 mM solution in DMSO.
  • Molecular weight 470.6 g/mol; chemical formula C31H34O4.
  • Suitable for in vitro pharmacology and receptor mechanism studies.