Learn More
Enterprise Technology Solutions Screen And Lens Cleaner Kit, 12 Oz Spray/Microfiber Cloth
SDP
Catalog No. 502041115
Encompass_Preferred
ABILITYONE 6850016831761 Skilcraft Safely clean glass and plastic screens and lenses with this streak-free anti-static cleaner. Safe for plasma and LCD screens and scientific and optical lenses, the ammonia- and alcohol-free formula removes dirt, grease, makeup, and fingerprints from all electronic devices. Kit includes one 12-ounce bottle, sprayer nozzle, and 12" x 12" microfiber cleaning cloth.
Catalog No. 50-204-1115 Supplier Enterprise Technology Solutions Supplier No. NSN6831761
ABILITYONE 6850016831761 Skilcraft Safely clean glass and plastic screens and lenses with this streak-free anti-static cleaner. Safe for plasma and LCD screens and scientific and optical lenses, the ammonia- and alcohol-free formula removes dirt, grease, makeup, and fingerprints from all electronic devices. Kit includes one 12-ounce bottle, sprayer nozzle, and 12" x 12" microfiber cleaning cloth.