Learn More
Sino Biological Recombinant Aequorea victoria GFP Protein (His Tag)
Supplier:  Sino Biological 69001S08E50
Encompass_Preferred
A DNA sequence encoding the Aequorea victoria GFP (AAB65663) (Met1-Leu238, except for the point mutations of intellectual property :F64L, S65T, R80K, S99F, T153M, I167V, H231L) was expressed, with a polyhistide tag at the C-terminus (Patent 201410599023.9).
Catalog No. 50-162-3271