Learn More
Sino Biological Recombinant Discosoma sp Orange fluorescent protein Protein (His Tag)
Supplier:  Sino Biological 69002S08E50
Encompass_Preferred
OFPSparkTM is a red (orange) fluorescent protein derived from DsRed(similarity is 98%). OFPSparkTM was expressed with a polyhistide tag at the C-terminus(Patent 201510003374.3).
Catalog No. 50-162-3272