Learn More
Sino Biological Recombinant Dermatophagoides pteronyssinus Der p 1 Protein (His Tag)
Supplier:  Sino Biological 69007A08H250
Encompass_Preferred
A DNA sequence encoding the dermatophagoides pteronyssinus DERP1 Protein was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3273