Learn More
Epredia™ Syntri Safeguard Tissue Processing Reagents
Catalog No. 6915ss Shop All Epredia Products
Change view
Click to view available options
Product Type:
10% Neutral Buffered Formalin
Clear-Rite 3
Dehydrant 100%
Fixation Buffer
Flex 100
Formal-Fixx
Isopropyl Alcohol
Tissue Processing Reagent
Xylene
Xylene Substitute
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
6915SS Clear-Rite 3
6764515SS Formal-Fixx
6615SS Xylene
6215SS Dehydrant 100%
8115SS Flex 100
9517SS Isopropyl Alcohol
5735SS 10% Neutral Buffered Formalin
6764215SS Xylene Substitute
61-15S-S Tissue Processing Reagent
61-15S-SS Fixation Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 6915SS Supplier Epredia™ Supplier No. 6915SS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tissue processing reagents that feature RFID technology for reagent management on the Epredia™ Revos™ Tissue Processor.
The Syntri Safeguard Reagents come in 5 liter bottles for tissue processing with RFID tags embedded in the labels. When partnered with the Epredia™ Revos™ Tissue Processor, Syntri Safeguard Reagent usage is tracked and logged on the instrument. The technology remembers every bottle loaded, documents every protocol executed and maintenance performed. Helping to reduce downtime and facilitates proper use of reagents.
Specifications
Product Type Clear-Rite 3
Quantity 4 x 5L
WARNING: Reproductive Harm - www.P65Warnings.ca.gov