Learn More
eMolecules​ 69739-34-0 | tert-Butyldimethylsilyl trifluoromethanesulfonate | Oakwood Chemicals | MFCD00000405 | 264.330 | C7H15F3O3SSi | 98.000 | CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F | 1g | 486548747
Catalog No. 5000170309 Shop All eMolecules Inc. Products
Encompass_Preferred
tert-Butyldimethylsilyl trifluoromethanesulfonate | Oakwood Chemicals | 69739-34-0 | MFCD00000405 | 264.330 | C7H15F3O3SSi | 98.000 | CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F | 1g | 486548747
Catalog No. 50-001-70309 Supplier eMolecules​ Supplier No. 486548747
tert-Butyldimethylsilyl trifluoromethanesulfonate | Oakwood Chemicals | 69739-34-0 | MFCD00000405 | 264.330 | C7H15F3O3SSi | 98.000 | CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F | 1g | 486548747