Learn More
Medchemexpress LLC 6A-azido-6A-deoxy-β-cyclodextrin | 98169-85-8 | MFCD05864973 | >85.0% | 1160.00 | C42H69N3O34 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10153225MG
Encompass_Preferred
6A-azido-6A-deoxy-β-cyclodextrin is an azide-functionalized derivative of β-cyclodextrin used as a water-soluble click-chemistry reagent and bioconjugation building block. The azido group enables copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions for conjugation to alkyne-bearing molecules in chemical biology and drug-delivery research.
  • Azide-functionalized for copper-catalyzed and strain-promoted azide-alkyne cycloaddition.
  • Water-soluble β-cyclodextrin derivative for aqueous reactions.
  • Suitable for bioconjugation and drug delivery research.
  • Enables site-specific conjugation to alkyne-containing partners.
  • Solid form, suitable for standard handling and storage.
  • Compatible with common analytical methods such as HPLC and NMR.
Catalog No. 50-002-45812