Learn More
Medchemexpress LLC 6A-azido-6A-deoxy-β-cyclodextrin | 98169-85-8 | MFCD05864973 | 98.0% | 1,160.00 | C42H69N3O34 | 5 MG
SDP
Catalog No. 5000248968
Encompass_Preferred
6A-azido-6A-deoxy-β-cyclodextrin is an azide-functionalized β-cyclodextrin derivative used as a reagent for click chemistry and bioconjugation. It contains a single azido substituent at the 6A position, is water-soluble, and enables efficient labeling and conjugation under aqueous conditions.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation.
  • Single azido handle for site-selective modification.
  • Water-soluble scaffold compatible with aqueous reactions.
  • Applicable to host-guest chemistry and carrier modification.
  • Supplied with high reported chemical purity for reproducible results.
Catalog No. 50-002-48968 Supplier Medchemexpress LLC Supplier No. HY1015325MG
6A-azido-6A-deoxy-β-cyclodextrin is an azide-functionalized β-cyclodextrin derivative used as a reagent for click chemistry and bioconjugation. It contains a single azido substituent at the 6A position, is water-soluble, and enables efficient labeling and conjugation under aqueous conditions.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation.
  • Single azido handle for site-selective modification.
  • Water-soluble scaffold compatible with aqueous reactions.
  • Applicable to host-guest chemistry and carrier modification.
  • Supplied with high reported chemical purity for reproducible results.