Learn More
Medchemexpress LLC 6A-azido-6A-deoxy-β-cyclodextrin | 98169-85-8 | MFCD05864973 | 98.0% | 1,160.00 g·mol⁻¹ | C42H69N3O34 | 10 MG
SDP
Catalog No. 5000246484
Encompass_Preferred
6A-Azido-6A-deoxy-β-cyclodextrin is a mono-azido functionalized β-cyclodextrin supplied as a solid. It provides an azide handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry) and is used for bioconjugation, drug-delivery research, and surface modification. High purity and defined molecular weight support analytical and synthetic workflows.
  • Mono-azido functionality for efficient click chemistry reactions.
  • High purity (98.0%) suitable for research applications.
  • Water-soluble cyclodextrin derivative for host-guest studies.
  • Good solubility in DMSO; ultrasonic assistance recommended.
  • Supplied as a solid for convenient handling and storage.
Catalog No. 50-002-46484 Supplier Medchemexpress LLC Supplier No. HY10153210MG
6A-Azido-6A-deoxy-β-cyclodextrin is a mono-azido functionalized β-cyclodextrin supplied as a solid. It provides an azide handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry) and is used for bioconjugation, drug-delivery research, and surface modification. High purity and defined molecular weight support analytical and synthetic workflows.
  • Mono-azido functionality for efficient click chemistry reactions.
  • High purity (98.0%) suitable for research applications.
  • Water-soluble cyclodextrin derivative for host-guest studies.
  • Good solubility in DMSO; ultrasonic assistance recommended.
  • Supplied as a solid for convenient handling and storage.