Learn More
Medchemexpress LLC 6A-azido-6A-deoxy-β-cyclodextrin | 98169-85-8 | MFCD05864973 | 98.0% | 1160.00 g·mol⁻1 | C42H69N3O34 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101532100MG
Encompass_Preferred
6A-Azido-6A-deoxy-β-cyclodextrin is an azide-functionalized β-cyclodextrin supplied as a solid reagent for click chemistry and bioconjugation. It offers a single azide handle on the cyclodextrin scaffold for site-selective conjugation and host-guest chemistry studies.
  • Azide functional group suitable for copper-catalyzed azide-alkyne cycloaddition.
  • High purity (98.0%) for reproducible synthetic and analytical work.
  • Good solubility in DMSO (≈125 mg/mL) for stock solution preparation.
  • Stable as powder at -20°C for long-term storage and in solution at -80°C for months.
  • Useful for bioconjugation, drug-delivery, and supramolecular chemistry applications.
Catalog No. 50-002-46483