Learn More
Medchemexpress LLC 6A-azido-6A-deoxy-β-cyclodextrin | 98169-85-8 | MFCD05864973 | 98.0% | 1160.00 g·mol⁻1 | C42H69N3O34 | 50 MG
SDP
Catalog No. 5000248004
Encompass_Preferred
6A-azido-6A-deoxy-β-cyclodextrin is a mono-substituted β-cyclodextrin bearing an azide functional group at the 6A position. It is a water-soluble derivative commonly used as a linker in click chemistry for bioconjugation and for host-guest complexation studies.
  • Contains an azide group for CuAAC and SPAAC click reactions.
  • High purity (98.0%).
  • Water-soluble mono-substituted β-cyclodextrin.
  • Suitable for bioconjugation and host-guest complexation studies.
  • Solid form for straightforward handling and weighing.
Catalog No. 50-002-48004 Supplier Medchemexpress LLC Supplier No. HY10153250MG
6A-azido-6A-deoxy-β-cyclodextrin is a mono-substituted β-cyclodextrin bearing an azide functional group at the 6A position. It is a water-soluble derivative commonly used as a linker in click chemistry for bioconjugation and for host-guest complexation studies.
  • Contains an azide group for CuAAC and SPAAC click reactions.
  • High purity (98.0%).
  • Water-soluble mono-substituted β-cyclodextrin.
  • Suitable for bioconjugation and host-guest complexation studies.
  • Solid form for straightforward handling and weighing.