Learn More
Medchemexpress LLC 6alpha-ethyl-3alpha,7alpha,23-trihydroxy-24-nor-5beta-cholan-23-sulfate sodium salt | 1000403-03-1 | 99.8% | 494.66 g/mol | C25H43NaO6S | 10MM 1ML
SDP
Catalog No. 5000236943
Encompass_Preferred
INT-767 is a research-grade dual farnesoid X receptor (FXR) and TGR5 agonist with reported mean EC50 values of 30 nM (FXR) and 630 nM (TGR5). It is supplied as a white to off-white solid and is available as a 10 mM solution in DMSO. Molecular formula C25H43NaO6S; molecular weight 494.66 g/mol. Intended for laboratory research use only.
  • Potent dual FXR and TGR5 agonist with EC50s of 30 nM and 630 nM, respectively.
  • High purity suitable for research applications.
  • Available as solid or 10 mM DMSO solution for convenience.
  • Highly soluble in DMSO and water according to manufacturer data.
  • Stable under recommended storage: solid at 4°C; in solution at -80°C for long-term storage.
  • Used in studies of bile acid signaling, metabolic disease, and liver injury models.
Catalog No. 50-002-36943 Supplier Medchemexpress LLC Supplier No. HY1243410MM 1ML
INT-767 is a research-grade dual farnesoid X receptor (FXR) and TGR5 agonist with reported mean EC50 values of 30 nM (FXR) and 630 nM (TGR5). It is supplied as a white to off-white solid and is available as a 10 mM solution in DMSO. Molecular formula C25H43NaO6S; molecular weight 494.66 g/mol. Intended for laboratory research use only.
  • Potent dual FXR and TGR5 agonist with EC50s of 30 nM and 630 nM, respectively.
  • High purity suitable for research applications.
  • Available as solid or 10 mM DMSO solution for convenience.
  • Highly soluble in DMSO and water according to manufacturer data.
  • Stable under recommended storage: solid at 4°C; in solution at -80°C for long-term storage.
  • Used in studies of bile acid signaling, metabolic disease, and liver injury models.