Learn More
Medchemexpress LLC 6alpha-ethyl-3alpha,7alpha,23-trihydroxy-24-nor-5beta-cholan-23-sulfate sodium salt | 1000403-03-1 | 98.0% | 494.7 g/mol | C25H43NaO6S | 1MG
SDP
Catalog No. 5000238496
Encompass_Preferred
INT-767 is a semisynthetic bile acid-derived small molecule used in preclinical research as a potent dual agonist of the farnesoid X receptor (FXR) and the G protein-coupled bile acid receptor TGR5. It modulates bile acid signaling and has been studied in liver and metabolic disease models in vitro and in vivo.
Catalog No. 50-002-38496 Supplier Medchemexpress LLC Supplier No. HY124341MG
INT-767 is a semisynthetic bile acid-derived small molecule used in preclinical research as a potent dual agonist of the farnesoid X receptor (FXR) and the G protein-coupled bile acid receptor TGR5. It modulates bile acid signaling and has been studied in liver and metabolic disease models in vitro and in vivo.