Learn More
Cayman Chemical 6amethyl11hydroxy ProgSt 100mg
SDP
Catalog No. 502982594
Encompass_Preferred
An anti-inflammatory steroid; a hydroxylated progesterone that lacks progestational and androgenic activity; inhibits phytohemagglutinin-activated T cell proliferation in vitro (IC50 = 2.9 μg/ml); inhibits TNF-α production in LPS-stimulated THP-1 cells (IC50 = 30.54 μg/ml)
Catalog No. 50-298-2594 Supplier Cayman Chemical Supplier No. 19533100 MG
An anti-inflammatory steroid; a hydroxylated progesterone that lacks progestational and androgenic activity; inhibits phytohemagglutinin-activated T cell proliferation in vitro (IC50 = 2.9 μg/ml); inhibits TNF-α production in LPS-stimulated THP-1 cells (IC50 = 30.54 μg/ml)