Learn More
Cayman Chemical 6α-Methylprednisolone 21-acetate, Purity: ≥95%, CAS: 53-36-1, 100 mg
SDP
Catalog No. 502913016
Encompass_Preferred
A synthetic glucocorticoid and an ester form of methylprednisolone; reduces IL-1β-induced production of PGE2 and increases in MMP-13 levels in equine osteochondral and synovial explant cocultures from 1-100 nM; intramuscular injection induces retinal lipid peroxidation and apoptosis in rabbits at 20 mg/kg
Catalog No. 50-291-3016 Supplier Cayman Chemical Supplier No. 35799100 MG
A synthetic glucocorticoid and an ester form of methylprednisolone; reduces IL-1β-induced production of PGE2 and increases in MMP-13 levels in equine osteochondral and synovial explant cocultures from 1-100 nM; intramuscular injection induces retinal lipid peroxidation and apoptosis in rabbits at 20 mg/kg