Learn More
Cayman Chemical 6aMethylprednIsoLN 21hemI 10mg
SDP
Supplier:  Cayman Chemical 1180010 MG
Encompass_Preferred
A prodrug form of methylprednisolone; nanoliposomes containing 6α-methylprednisolone 21-hemisuccinate decrease disease severity in a rat model of adjuvant-induced arthritis
Catalog No. 50-295-1995