Learn More
Cayman Chemical 6α-Methylprednisolone 21-hemisuccinate (sodium salt), Purity: ≥95%, CAS: 2375-03-3, 50 mg
SDP
Catalog No. 502910448
Encompass_Preferred
A prodrug form of methylprednisolone; nanoliposomes containing 6α-methylprednisolone 21-hemisuccinate decrease disease severity in a rat model of adjuvant-induced arthritis
Catalog No. 50-291-0448 Supplier Cayman Chemical Supplier No. 3574250 MG
A prodrug form of methylprednisolone; nanoliposomes containing 6α-methylprednisolone 21-hemisuccinate decrease disease severity in a rat model of adjuvant-induced arthritis