Learn More
Cayman Chemical 6AmIno8trIfluoromthylphen 10mg
SDP
Supplier:  Cayman Chemical 2641910 MG
Encompass_Preferred
An antiprion agent; inhibits protein folding activity of the ribosome at 150 µM; directly competes with protein substrates for the ribosomal active site
Catalog No. 50-293-3989