Learn More
AdipoGen 6BIO (10 mg)
SDP
Supplier:  AdipoGen AGMRC0019M010
Encompass_Preferred
Chemical. CAS: 667463-62-9. Formula: C16H10BrN3O2. MW: 356.2. Potent, reversible and ATP-competitive glycogen synthase kinase-3alpha/beta (GSK-3alpha/beta) inhibitor. JAK/STAT3 signaling inhibitor. Phosphoinositide-dependent kinase 1 (PDK1) inhibitor. Anticancer compound. Potent antiproliferative agent. Suppresses metastasis. Apoptosis inducer. Sustains pluripotency and self-renewal of human and mouse embryonic stem cells (ESCs) by activation of the Wnt signaling pathway. Anti-leishmanial (IC50 = 0.150µM). Proto-oncogene tyrosine-protein kinase receptor RET inhibitor (IC50=0.51µM). Inhibits HIV-1 transcription and protects against Tat induced neurotoxicity.
Catalog No. 50-205-4238